missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl isocyanate, 95%

Product Code. 11498097
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498097 100 g 100g
11474000 25 g 25g
11408107 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498097 Supplier Thermo Scientific Alfa Aesar Supplier No. A17492.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4083-64-1 | C8H7NO3S | 197.21 g/mol

It is a reagent used to prepare acetylated syn-1,2-diols,oxazolidin-2-ones, 2,3-diamino acids, and N-tosylcarbonamides. -Toluenesulfonyl isocyanate has been used as derivatization reagent in determination of 3-α-hydroxy tibolone in human plasma by LC-MS/MS. It is also employed in the derivatization reagent in simultaneous quantitative analysis of diethylene glycol and propylene glycol in pharmaceutical products by HPLC and as reagent in the preparation of acetylated syn-1,2-diols, oxazolidin-2-ones, 2,3-diamino acids and N-tosylcarbonamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent used to prepare acetylated syn-1,2-diols,oxazolidin-2-ones, 2,3-diamino acids, and N-tosylcarbonamides. -Toluenesulfonyl isocyanate has been used as derivatization reagent in determination of 3-α-hydroxy tibolone in human plasma by LC-MS/MS. It is also employed in the derivatization reagent in simultaneous quantitative analysis of diethylene glycol and propylene glycol in pharmaceutical products by HPLC and as reagent in the preparation of acetylated syn-1,2-diols, oxazolidin-2-ones, 2,3-diamino acids and N-tosylcarbonamides.

Solubility
Reacts with water.

Notes
Keep away from alcohols, strong bases, amines, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4083-64-1
Molecular Formula C8H7NO3S
Molecular Weight (g/mol) 197.21
MDL Number MFCD00002030
InChI Key VLJQDHDVZJXNQL-UHFFFAOYSA-N
Synonym tosyl isocyanate, p-toluenesulfonyl isocyanate, 4-methylbenzenesulfonyl isocyanate, p-toluenesulphonyl isocyanate, benzenesulfonyl isocyanate, 4-methyl, tosylisocyanate, p-tolylsulfonyl isocyanate, unii-h9004fjx6v, p-toluenesulfonic acid, anhydride with isocyanic acid, p-toluene sulfonyl isocyanate
PubChem CID 77703
IUPAC Name 4-methyl-N-(oxomethylidene)benzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N=C=O

Specifications

Melting Point 5°C
Density 1.295
Boiling Point 144°C (10 mmHg)
Flash Point 145°C (293°F)
Odor Irritating
Refractive Index 1.535
Quantity 100 g
UN Number UN2206
Beilstein 391287
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 197.21
Percent Purity 95%
Chemical Name or Material p-Toluenesulfonyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.