missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl fluoride, 97%, Thermo Scientific™

Product Code. 10376401 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10376401 25 g Glass Bottle 25g
10438872 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10376401 Supplier Thermo Scientific Acros Supplier No. 152380250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-16-3
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
InChI Key IZZYABADQVQHLC-UHFFFAOYSA-N
Synonym p-toluenesulfonyl fluoride, tosyl fluoride, 4-toluenesulfonyl fluoride, benzenesulfonyl fluoride, 4-methyl, 4-tolylsulfonyl fluoride, toluene-4-sulfonyl fluoride, p-methylbenzenesulfonyl fluoride, p-toluenesulphonyl fluoride, toluene-p-sulphonyl fluoride, p-methylphenylsulfonyl fluoride
PubChem CID 9965
IUPAC Name 4-methylbenzenesulfonyl fluoride
SMILES CC1=CC=C(C=C1)S(=O)(=O)F

Specifications

Melting Point 40.0°C to 43.0°C
Boiling Point 112.0°C (16.0 mmHg)
Flash Point 106°C
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula CH3C6H4SO2F
Quantity 25 g
Beilstein 11, II, 54
Solubility Information Solubility in water: hydrolysis.
Formula Weight 174.19
Percent Purity 97%
Chemical Name or Material p-Toluenesulfonyl fluoride, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Toluenesulfonyl fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.