missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
Melting Point | 67.0°C to 69.0°C |
Color | White |
Boiling Point | 134.0°C (10.0 mmHg) |
Flash Point | 128°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min |
Packaging | Glass bottle |
Linear Formula | CH3C6H4SO2Cl |
Quantity | 500 g |
Beilstein | 11,103 |
Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- p-Toluenesulfonyl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'