missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic acid zinc salt hydrate

Product Code. 11377977
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377977 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11377977 Supplier Thermo Scientific Alfa Aesar Supplier No. 020899.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 123334-05-4 | C14H14O6S2Zn | 407.76 g/mol

p-Toluenesulfonic acid zinc salt hydrate is used as a zinc benzenesulfonate salt for proteomics research, and as s a stabilizer & lubricants & activator, it's used in synthetic rubber of PE, PVC, polypropylene nylon, ABS natural rubber, butylbenzene rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123334-05-4
Molecular Formula C14H14O6S2Zn
Molecular Weight (g/mol) 407.76
MDL Number MFCD00151013
InChI Key YISPIDBWTUCKKH-UHFFFAOYSA-L
Synonym zinc p-toluenesulfonate hydrate, acmc-20eh0l, p-toluenesulfonic acid zinc salt, zinc 2+ hydrate ditosylate, p-toluenesulfonic acid zinc salt hydrate, benzenesulfonic acid,4-methyl-, zinc salt, hydrate 2:1:?
PubChem CID 16212267
IUPAC Name zinc;4-methylbenzenesulfonate;hydrate
SMILES [Zn++].CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 407.76
Physical Form Powder
Chemical Name or Material p-Toluenesulfonic acid zinc salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.