missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals

Catalog Number 10286913
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
6192-52-5
MFCD00142137
KJIFKLIQANRMOU-UHFFFAOYSA-N
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
521998
4-methylbenzenesulfonic acid;hydrate
CC1=CC=C(C=C1)S(=O)(=O)O.O
This item is not returnable. View return policy
6192-52-5
MFCD00142137
KJIFKLIQANRMOU-UHFFFAOYSA-N
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
521998
4-methylbenzenesulfonic acid;hydrate
CC1=CC=C(C=C1)S(=O)(=O)O.O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100.0°C to 108.0°C
Color Pink to White
Flash Point 180°C
Infrared Spectrum Authentic
Assay Percent Range 97.5%
Packaging Plastic drum
Molecular Formula C7H10O4S
Linear Formula CH3C6H4SO3H·H2O
Quantity 5 kg
Beilstein 11, 97
Fieser 01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507
Merck Index 15, 9692
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and chloroform
Molecular Weight (g/mol) 190.213
Formula Weight 190.22
Percent Purity 97.5%
Grade Pure
Physical Form Agglomerating Crystals
Chemical Name or Material p-Toluenesulfonic acid monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • p-Toluenesulfonic acid monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'