missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfinic acid sodium salt, 97% (dry wt.), water <5%

Product Code. 11400283
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400283 100 g 100g
11404010 25 g 25g
11410283 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400283 Supplier Thermo Scientific Alfa Aesar Supplier No. A11430.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 824-79-3 | C7H7NaO2S | 178.181 g/mol

p-Toluenesulfinic acid sodium salt is used as an intermediate for the preparation of pharmaceuticals and dispersal dyes. It acts as a curing agent of grouting material. It is also used as a discharge medium of dyes and pigments. Further, it serves as a chemical intermediate in the production of fluorescent pigments and fabric adhesive. In addition to this, it is used as an electroplating brightening agent and curing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfinic acid sodium salt is used as an intermediate for the preparation of pharmaceuticals and dispersal dyes. It acts as a curing agent of grouting material. It is also used as a discharge medium of dyes and pigments. Further, it serves as a chemical intermediate in the production of fluorescent pigments and fabric adhesive. In addition to this, it is used as an electroplating brightening agent and curing agent.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-79-3
Molecular Formula C7H7NaO2S
Molecular Weight (g/mol) 178.181
MDL Number MFCD00149640
InChI Key KFZUDNZQQCWGKF-UHFFFAOYSA-M
Synonym sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech
PubChem CID 2723791
IUPAC Name sodium;4-methylbenzenesulfinate
SMILES CC1=CC=C(C=C1)S(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 4621454
Solubility Information Slightly soluble in water.
Formula Weight 178.19
Percent Purity 97%
Assay (dry wt.)
Chemical Name or Material p-Toluenesulfinic acid sodium salt, Water <5%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.