missing translation for 'onlineSavingsMsg'
Learn More

p-Toluamide, 98+%

Product Code. 11464603
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464603 100 g 100g
11454603 25 g 25g
11474603 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464603 Supplier Thermo Scientific Alfa Aesar Supplier No. A11559.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-55-6 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-55-6
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00008002
InChI Key UHBGYFCCKRAEHA-UHFFFAOYSA-N
Synonym p-toluamide, benzamide, 4-methyl, p-methylbenzamide, 4-methyl-benzamide, 4-toluamide, 4-carbamoyltoluene, unii-n0v326v2f5, tolylsaureamid, n-toluamide, p-tolylamide
PubChem CID 69274
IUPAC Name 4-methylbenzamide
SMILES CC1=CC=C(C=C1)C(N)=O

Specifications

Melting Point 158°C to 160°C
Quantity 100 g
Beilstein 774353
Formula Weight 135.17
Percent Purity ≥98%
Chemical Name or Material p-Toluamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.