missing translation for 'onlineSavingsMsg'
Learn More

p-Tolualdehyde, 98%, Thermo Scientific Chemicals

£18.20 - £349.00

Chemical Identifiers

CAS 104-87-0
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00006954
InChI Key FXLOVSHXALFLKQ-UHFFFAOYSA-N
Synonym p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
PubChem CID 7725
ChEBI CHEBI:28617
IUPAC Name 4-methylbenzaldehyde
SMILES CC1=CC=C(C=C1)C=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11414010
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11384.06
5 g
£18.20
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11410213
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11384.22
100 g
£26.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11420213
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11384.36
500 g
£89.80
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11430213
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11384.0E
2500 g
£349.00
2500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

Solubility
It is slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Incompatible to Strong bases, Strong oxidizing agents, Strong reducing agents
Specifications

Chemical Identifiers

104-87-0
120.151
FXLOVSHXALFLKQ-UHFFFAOYSA-N
7725
4-methylbenzaldehyde
C8H8O
MFCD00006954
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
CHEBI:28617
CC1=CC=C(C=C1)C=O

Specifications

104-87-0
1.018
80°C (176°F)
1.546
385772
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
FXLOVSHXALFLKQ-UHFFFAOYSA-N
4-methylbenzaldehyde
7725
120.15
p-Tolualdehyde
-6°C
204°C
C8H8O
MFCD00006954
Air Sensitive
It is slightly soluble in water.
CC1=CC=C(C=C1)C=O
120.151
CHEBI:28617
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319-H335

missing translation for 'einecsNumber' : 203-246-9

missing translation for 'rtecsNumber' : CU7034500

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only