missing translation for 'onlineSavingsMsg'
Learn More

p-Tolualdehyde, 98%, Thermo Scientific Chemicals

Catalog Number 11430213
Click to view available options
Quantity:
5 g
100 g
500 g
2500 g
This item is not returnable. View return policy
104-87-0
C8H8O
120.151
MFCD00006954
FXLOVSHXALFLKQ-UHFFFAOYSA-N
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
7725
CHEBI:28617
4-methylbenzaldehyde
CC1=CC=C(C=C1)C=O
This item is not returnable. View return policy
104-87-0
C8H8O
120.151
MFCD00006954
FXLOVSHXALFLKQ-UHFFFAOYSA-N
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
7725
CHEBI:28617
4-methylbenzaldehyde
CC1=CC=C(C=C1)C=O

p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

Solubility
It is slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Incompatible to Strong bases, Strong oxidizing agents, Strong reducing agents
TRUSTED_SUSTAINABILITY
Melting Point -6°C
Density 1.018
Boiling Point 204°C
Flash Point 80°C (176°F)
Refractive Index 1.546
Quantity 2500 g
Beilstein 385772
Sensitivity Air Sensitive
Solubility Information It is slightly soluble in water.
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material p-Tolualdehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'