missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Terphenyl-d14, TRC
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
Specifications
Specifications
| Chemical Name or Material | p-Terphenyl D14 |
| CAS | 1718-51-0 |
| Molecular Formula | C18 2H14 |
| Quantity | 1 g |
| Synonym | 1,1':4',1''-Terphenyl-2,2',2'',3,3',3'',4,4'',5,5',5'',6,6',6''-d14, p-Terphenyl-d14 (7CI,8CI), 4-Terphenyl-d14, p-Terphenyl D14 |
| InChI Formula | InChI=1S/C18H14/c1-3-7-15(8-4-1)17-11-13-18(14-12-17)16-9-5-2-6-10-16/h1-14H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D |
| SMILES | [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c([2H])c([2H])c(c([2H])c2[2H])c3c([2H])c([2H])c([2H])c([2H])c3[2H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-[2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)phenyl]benzene |
| Molecular Weight (g/mol) | 244.39 |
| Formula Weight | 244.1974 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction