missing translation for 'onlineSavingsMsg'
Learn More

p-Terphenyl-d14, TRC

Product Code. 30424076
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30424076

Brand: Toronto Research Chemicals TRCT116682500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Terphenyl D14
CAS 1718-51-0
Molecular Formula C18 2H14
Quantity 500 mg
Synonym 1,1':4',1''-Terphenyl-2,2',2'',3,3',3'',4,4'',5,5',5'',6,6',6''-d14, p-Terphenyl-d14 (7CI,8CI), 4-Terphenyl-d14, p-Terphenyl D14
InChI Formula InChI=1S/C18H14/c1-3-7-15(8-4-1)17-11-13-18(14-12-17)16-9-5-2-6-10-16/h1-14H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D
SMILES [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c([2H])c([2H])c(c([2H])c2[2H])c3c([2H])c([2H])c([2H])c([2H])c3[2H]
IUPAC Name 1,2,3,4,5-pentadeuterio-6-[2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)phenyl]benzene
Molecular Weight (g/mol) 244.39
Formula Weight 244.1974
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.