missing translation for 'onlineSavingsMsg'
Learn More

p-Terphenyl D14 in DCM 2000μg/mL, Fisher chemical™

Product Code. 12997103 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Ampoule
Bottle
Unit Size:
1mL
5mL
This item is not returnable. View return policy

Product Code. 12997103

Brand: Fisher Chemical P887010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 1718-51-0
Molecular Formula C18H14
Molecular Weight (g/mol) 244.395
InChI Key XJKSTNDFUHDPQJ-WZAAGXFHSA-N
PubChem CID 123128
IUPAC Name 1,2,3,4,5-pentadeuterio-6-[2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)phenyl]benzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3

Specifications

Packaging Bottle
Analyte or Component Names p-Terphenyl D14
Concentration 2000 μg/mL
Type Pesticide and Metabolite Standards
Element p-Terphenyl D14
Solvent or Matrix Dichloromethane
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.