missing translation for 'onlineSavingsMsg'
Learn More

p-Terphenyl D14 500 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30325265
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30325265

Brand: Dr. Ehrenstorfer DREGA09010298DI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Terphenyl D14
CAS 1718-51-0
Solvent or Matrix Dichloromethane
Molecular Formula C18 2H14
Quantity 1 mL
Synonym 1,1':4',1''-Terphenyl-2,2',2'',3,3',3'',4,4'',5,5',5'',6,6',6''-d14, p-Terphenyl-d14 (7CI,8CI), 4-Terphenyl-d14, p-Terphenyl D14
InChI Formula InChI=1S/C18H14/c1-3-7-15(8-4-1)17-11-13-18(14-12-17)16-9-5-2-6-10-16/h1-14H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D
SMILES [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c([2H])c([2H])c(c([2H])c2[2H])c3c([2H])c([2H])c([2H])c([2H])c3[2H]
IUPAC Name 1,2,3,4,5-pentadeuterio-6-[2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)phenyl]benzene
Molecular Weight (g/mol) 244.39
Formula Weight 244.1974 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.