missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ P-Selectin/CD62P Antibody (03) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30734MFV500
This item is not returnable.
View return policy
Description
P-Selectin/CD62P Monoclonal antibody specifically detects P-Selectin/CD62P in Human samples. It is validated for ELISA
Specifications
| P-Selectin/CD62P | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
| This product is a recombinant monoclonal antibody expressed from HEK293 cells made from recombinant Human P-Selectin/CD62P protein (RefSeq#: AAN06828.1) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 03 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6403 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction