missing translation for 'onlineSavingsMsg'
Learn More

P-Selectin/CD62P Antibody (001), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18418608 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418608 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418608 Supplier Novus Biologicals Supplier No. NBP290703JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

P-Selectin/CD62P Monoclonal antibody specifically detects P-Selectin/CD62P in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Host Species Rabbit
Purification Method Protein A purified
Quantity 0.1 mL
Gene ID (Entrez) 6403
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.