missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | P-Selectin/CD62P |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1 |
| Conjugate | Alexa Fluor 594 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human P-Selectin/CD62P (AAN06828.1; Met1-Ala771). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?