missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Rex1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97481AF594
This item is not returnable.
View return policy
Description
P-Rex1 Polyclonal specifically detects P-Rex1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| P-Rex1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| KIAA1415, phosphatidylinositol 34,5-trisphosphate-dependent Rac exchanger 1 protein, phosphatidylinositol-34,5-trisphosphate-dependent Rac exchange factor 1, ptdIns(34,5)-dependent Rac exchanger 1 | |
| Produced in rabbits immunized with E. coli-derived Human P-Rex1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 57580 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction