missing translation for 'onlineSavingsMsg'
Learn More

P-Rex1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18353092 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353092 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353092 Supplier Novus Biologicals Supplier No. NBP298883AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

P-Rex1 Polyclonal specifically detects P-Rex1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P-Rex1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA1415, phosphatidylinositol 34,5-trisphosphate-dependent Rac exchanger 1 protein, phosphatidylinositol-34,5-trisphosphate-dependent Rac exchange factor 1, ptdIns(34,5)-dependent Rac exchanger 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human P-Rex1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57580
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.