missing translation for 'onlineSavingsMsg'
Learn More

p-Quinquephenyl, 98%, Thermo Scientific Chemicals

Product Code. p-7028893
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15423447

Brand: Thermo Scientific Alfa Aesar H26308.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3073-05-0
Molecular Formula C30H22
Molecular Weight (g/mol) 382.506
MDL Number MFCD00059012
InChI Key OMCUOJTVNIHQTI-UHFFFAOYSA-N
Synonym p-quinquephenyl, p-pentaphenyl, 1,1':4',1:4,1':4',1-quinquephenyl, p-quinquiphenyl, acmc-209hie, 1,4-bis 4-phenylphenyl benzene, 1-phenyl-4-4-4-phenylphenyl phenyl benzene, 1,1:4,1':4',1-quinquephenyl, 4-1,1'-biphenyl-4-yl-4'-phenyl-1,1'-biphenyl
PubChem CID 137813
IUPAC Name 1,4-bis(4-phenylphenyl)benzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=CC=C5

Specifications

Quantity 250 mg
Formula Weight 382.5
Percent Purity 98%
Chemical Name or Material p-Quinquephenyl

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.