missing translation for 'onlineSavingsMsg'
Learn More

p-Phenylenediamine sulfate, 98%, Thermo Scientific™

Product Code. 11460523
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460523 250 g 250g
11450523 1000 g 1000g
11470523 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460523 Supplier Thermo Scientific Alfa Aesar Supplier No. A12659.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16245-77-5
Molecular Formula C6H10N2O4S
Molecular Weight (g/mol) 206.22
MDL Number MFCD00035510
InChI Key UFPKLWVNKAMAPE-UHFFFAOYSA-N
Synonym p-phenylenediamine sulfate, benzene-1,4-diamine sulfate, 1,4-diaminobenzene sulfate, 1,4-phenylenediamine sulfate, 1,4-benzenediamine, sulfate 1:1, unii-s7igm30961, benzene-1,4-diamine; sulfuric acid, 1,4-benzenediamine, sulfate, benzene-1,4-diammonium sulphate, 1,4-benzenediamine sulfate
PubChem CID 27769
IUPAC Name benzene-1,4-diamine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=CC=C(N)C=C1

Specifications

Melting Point >300°C
Quantity 250 g
UN Number UN2811
Formula Weight 206.22
Percent Purity 98%
Chemical Name or Material p-Phenylenediamine sulfate
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.