missing translation for 'onlineSavingsMsg'
Learn More

P-Phenylenediamine Dihydrochloride, TRC

Product Code. 30133206
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133206 2.5 g 2.5g
30012128 500 mg 500mg
30006488 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133206 Supplier Toronto Research Chemicals Supplier No. TRCP3996182.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 624-18-0
Molecular Formula C6H8N2 . 2HCl
Molecular Weight (g/mol) 181.06
Synonym 1,4-Diaminobenzene Dihydrochloride, 1,4-Phenylenediamine Dihydrochloride, C.I. 76061, C.I. Oxidation Base 10A, Durafur Black RC, Fourrine 64, Fourrine DS, Pelagol CD, Pelagol Grey CD;
IUPAC Name benzene-1,4-diamine;dihydrochloride
SMILES Cl.Cl.Nc1ccc(N)cc1

Specifikationer

Quantity 2.5 g
InChI Formula InChI=1S/C6H8N2.2ClH/c7-5-1-2-6(8)4-3-5;;/h1-4H,7-8H2;2*1H
Formula Weight 180.0221
Chemical Name or Material P-Phenylenediamine Dihydrochloride
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.