missing translation for 'onlineSavingsMsg'
Learn More

p-Phenylenediamine dihydrochloride, 99%, Thermo Scientific™

Product Code. 11483364
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483364 50 g 50g
11493364 250 g 250g
11473364 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483364 Supplier Thermo Scientific Alfa Aesar Supplier No. A10638.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-18-0
Molecular Formula C6H10Cl2N2
Molecular Weight (g/mol) 181.06
MDL Number MFCD00013002
InChI Key IYXMNTLBLQNMLM-UHFFFAOYSA-N
Synonym benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64
PubChem CID 12205
IUPAC Name benzene-1,4-diamine;dihydrochloride
SMILES C1=CC(=CC=C1N)N.Cl.Cl

Specifications

Melting Point ∼265°C
Quantity 50 g
UN Number UN2811
Beilstein 3911749
Merck Index 14,7285
Formula Weight 181.07
Percent Purity 99%
Chemical Name or Material p-Phenylenediamine dihydrochloride
compliance-icons compliance-icons
Product Identifier
  • p-Phenylenediamine dihydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P361-Take off immediately all contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.