missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Nitrophenyl Phosphate (Disodium Salt, Hexahydrate), Fisher BioReagents
Description
p-Nitrophenyl Phosphate is suitable for use as a substrate for alkaline and acid phosphatase.
Specifications
Specifications
| Melting Point | 300°C |
| Color | White |
| Assay Percent Range | ≥98.0% |
| Packaging | Amber Glass |
| Quantity | 5 g |
| Formula Weight | 371 |
| Physical Form | Powder or Solid |
| Chemical Name or Material | p-Nitrophenyl Phosphate Disodium Salt, Hexahydrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?