Learn More
Melford p-Nitrophenyl-N-acetyl-B-D-galactosaminide
Product Code. p-200087189
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
Product Code. 30018602
Brand: Melford N660400.25
Chromogenic substrate for detecting N-Acetyl-B-D-glucosaminidase.
Chromogenic substrate for detecting N-Acetyl-B-D-glucosaminidase.
Specifications
Quantity 250 mg