missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Nitrophenyl a-D-Glucopyranoside, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | p-Nitrophenyl a-D-Glucopyranoside |
| CAS | 3767-28-0 |
| Molecular Formula | C12 H15 N O8 |
| Quantity | 5 g |
| Synonym | α-D-Glucopyranoside','4-nitrophenyl, Glucopyranoside','p-nitrophenyl','α-D- (8CI), α-D-Glucopyranoside','p-nitrophenyl (7CI), α-D-Glucoside','p-nitrophenyl (6CI), 4-Nitrophenyl α-D-glucopyranoside, 4-Nitrophenyl α-D-glucoside, 4'-Nitrophenyl α-D-glucopyranoside, p-Nitrophenol α-D-glucoside, p-Nitrophenyl α-D-glucopyranoside, p-Nitrophenyl α-D-glucoside |
| InChI Formula | InChI=1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-3-1-6(2-4-7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9-,10+,11-,12+/m1/s1 |
| SMILES | OC[C@H]1O[C@H](Oc2ccc(cc2)[N+](=O)[O-])[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 301.25 |
| Formula Weight | 301.0798 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction