missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl 2-(Furfurylsulfinyl)acetate, TRC

Product Code. 30180401
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180401 50 g 50g
30179749 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180401 Supplier Toronto Research Chemicals Supplier No. TRCN50356550G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123855-55-0
Molecular Formula C13H11NO6S
Molecular Weight (g/mol) 309.29
Synonym 2-[(2-Furanylmethyl)sulfinyl]acetic Acid 4-Nitrophenyl Ester, [(2-Furanylmethyl)sulfinyl]acetic Acid 4-Nitrophenyl Ester;
IUPAC Name (4-nitrophenyl) 2-(furan-2-ylmethylsulfinyl)acetate
SMILES [O-][N+](=O)c1ccc(OC(=O)CS(=O)Cc2occc2)cc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C13H11NO6S/c15-13(9-21(18)8-12-2-1-7-19-12)20-11-5-3-10(4-6-11)14(16)17/h1-7H,8-9H2
Formula Weight 309.0307
Chemical Name or Material p-Nitrophenyl 2-(Furfurylsulfinyl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.