missing translation for 'onlineSavingsMsg'
Learn More

p-Methylstyrene, 97%, stabilized

Product Code. 10458152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
10mL
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10458152 500 mL Glass bottle 500mL
10357200 10 mL Glass bottle 10mL
10353812 100 mL Glass bottle 100mL
11914461 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10458152 Supplier Thermo Scientific Acros Supplier No. 127705000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-97-9 | C9H10 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-97-9
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00008621
InChI Key JLBJTVDPSNHSKJ-UHFFFAOYSA-N
Synonym 4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene
PubChem CID 12161
IUPAC Name 1-ethenyl-4-methylbenzene
SMILES CC1=CC=C(C=C)C=C1

Specifications

Melting Point -34.0°C
Color Colorless to Yellow
Density 0.8960g/mL
Boiling Point 170.0°C to 175.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH=CH2
Refractive Index 1.5410 to 1.5430
Quantity 500 mL
Beilstein 05,485
Specific Gravity 0.896
Solubility Information Solubility in water: insoluble
Stabilizer 0.1% 3, 5-di-tert.-butylcatechol
Formula Weight 118.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material p-Methylstyrene
Show More Show Less
compliance-icons
Product Identifier
  • p-Methylstyrene, stabilized
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.