missing translation for 'onlineSavingsMsg'
Learn More

P-Methylphosphonic Acid-13C, TRC

Product Code. 30482204
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482204 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482204 Supplier Toronto Research Chemicals Supplier No. TRCM32690750MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material P-Methylphosphonic Acid-13C
CAS 106976-96-9
Molecular Formula 13CH5O3P
Quantity 50 mg
Synonym Dihydrogen Methylphosphonate-13C, MPA-13C, Methanephosphonic Acid-13C, Methylphosphonic Acid-13C
InChI Formula InChI=1S/CH5O3P/c1-5(2,3)4/h1H3,(H2,2,3,4)/i1+1
SMILES O=P(O)(O)[13CH3]
Molecular Weight (g/mol) 97.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.