missing translation for 'onlineSavingsMsg'
Learn More

p-Mentha-8-thiol-3-one, cis + trans, 97%

Product Code. 11455956
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11455956

Brand: Thermo Scientific Alfa Aesar B22287.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38462-22-5 | C10H18OS | 186.313 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38462-22-5
Molecular Formula C10H18OS
Molecular Weight (g/mol) 186.313
MDL Number MFCD00012393
InChI Key RVOKNSFEAOYULQ-UHFFFAOYSA-N
Synonym p-mentha-8-thiol-3-one, 8-mercaptomenthone, 8-mercapto-p-menthan-3-one, thiomenthone, 8-mercapto-p-menthane-3-one, cyclohexanone, 2-1-mercapto-1-methylethyl-5-methyl, 2-1-mercapto-1-methylethyl-5-methylcyclohexanone, buchu mercaptan, 5-methyl-2-2-sulfanylpropan-2-yl cyclohexan-1-one, 2-1-mercapto-1-methylethyl-5-methylcyclohexan-1-one
PubChem CID 61982
IUPAC Name 5-methyl-2-(2-sulfanylpropan-2-yl)cyclohexan-1-one
SMILES CC1CCC(C(=O)C1)C(C)(C)S

Specifications

Density 1
Boiling Point 270°C to 272°C
Flash Point 108°C (226°F)
Quantity 5 g
Formula Weight 186.31
Percent Purity 97%
Chemical Name or Material p-Mentha-8-thiol-3-one, cis + trans
compliance-icons compliance-icons
Product Identifier
  • p-Mentha-8-thiol-3-one
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : p-Mentha-8-thiol-3-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.