missing translation for 'onlineSavingsMsg'
Learn More

p-Hydroxymephentermine Hydrochloride, TRC

Product Code. 30402325
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402325 10 mg
30407363 1 mg
30389574 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402325 Supplier Toronto Research Chemicals Supplier No. TRCH79172010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120583-18-8
Molecular Formula C11 H17 N O . Cl H
Molecular Weight (g/mol) 215.72
Synonym p-(2-Methyl-2-methylaminopropyl)phenol Hydrochloride, 4-[2-Methyl-2-(methylamino)propyl]phenol Hydrochloride
IUPAC Name 4-[2-methyl-2-(methylamino)propyl]phenol;hydrochloride
SMILES Cl.CNC(C)(C)Cc1ccc(O)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H17NO.ClH/c1-11(2,12-3)8-9-4-6-10(13)7-5-9;/h4-7,12-13H,8H2,1-3H3;1H
Formula Weight 215.1077
Chemical Name or Material p-Hydroxymephentermine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.