missing translation for 'onlineSavingsMsg'
Learn More

P-Hydroxybenzoic Acid Crystalline MP Biomedicals

Product Code. 11498200 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498200 1 kg 1kg
11478200 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498200 Supplier MP Biomedicals Supplier No. 0210257791
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 99-96-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002547
InChI Key FJKROLUGYXJWQN-UHFFFAOYSA-N
Synonym p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german
PubChem CID 135
ChEBI CHEBI:30763
IUPAC Name 4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C=C1

Specifications

Density 1.46 g/mL
Quantity 1 kg
Formula Weight 138.1
Physical Form Crystalline
Chemical Name or Material P-Hydroxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.