Learn More
Invitrogen™ P-Glycoprotein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532648
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%); Monkey (94%); Mouse, Hamster (82%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.
Specifications
| P-Glycoprotein | |
| Polyclonal | |
| Unconjugated | |
| ABCB1 | |
| ABC superfamily; P-glycoprotein; ABC20; ABCB1; Abcb1a; Abcb1b; Abcb4; ATP binding cassette subfamily B member 1; ATP-binding cassette sub-family B (MDR/TAP) member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette sub-family B member 1; ATP-binding cassette sub-family B member 1A; ATP-binding cassette sub-family B member 1B; ATP-binding cassette transporter subfamily B, member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette, subfamily B (MDR/TAP), member 1A; ATP-binding cassette, sub-family B (MDR/TAP), member 1A; ATP-binding cassette, subfamily B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 4; ATP-binding cassette, subfamily B, member 1A; ATP-binding cassette, subfamily B, member 1B; ATP-dependent translocase ABCB1; ATP-dependent translocase ABCB1; multidrug resistance protein 1; bovine p-glycoprotein; CD243; CLCS; colchicin sensitivity; doxorubicin resistance; ecotropic viral integration site 32; Evi32; GP170; mdr; Mdr1; MDR1A; Mdr1b; Mdr3; mdr-3; multi-drug resistance 3; multidrug resistance p-glycoprotein; multidrug resistance protein 1; Multidrug resistance protein 1A; multidrug resistance protein 1B; Multidrug resistance protein 2; multidrug resistance protein 3; multidrug resistance protein MDR1; multiple drug resistant 1a; P glycoprotein 1; P glycoprotein 3; P-glycoprotein; P-glycoprotein 1; P-glycoprotein 2; P-glycoprotein 3; P-glycoprotein/multidrug resistance 1; Pgp; p-gp; PGP1; PGP2; PGY1; Pgy-1; Pgy1-1; PGY2; Pgy3; Pgy-3; Phospholipid transporter ABCB1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5243 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 0.1% sodium azide | |
| P08183 | |
| ABCB1 | |
| Synthetic 17 amino acid peptide from cytoplasmic domain of human ABCB1 / MDR1. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.