missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P-Glycoprotein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119958
This item is not returnable.
View return policy
Description
Positive Control: HepG2 cell lysate, Hela cell lysate, human kidney tissue, human small intestine tissue, human thyroid carcinoma tissue, mouse kidney tissue, rat brain tissue. Subcellular Location: Cell membrane, Apical cell membrane.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.
Specifications
| P-Glycoprotein | |
| Polyclonal | |
| Unconjugated | |
| ABCB1 | |
| ABC superfamily; P-glycoprotein; ABC20; ABCB1; Abcb1a; Abcb1b; Abcb4; ATP binding cassette subfamily B member 1; ATP-binding cassette sub-family B (MDR/TAP) member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette sub-family B member 1; ATP-binding cassette sub-family B member 1A; ATP-binding cassette sub-family B member 1B; ATP-binding cassette transporter subfamily B, member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette, subfamily B (MDR/TAP), member 1A; ATP-binding cassette, sub-family B (MDR/TAP), member 1A; ATP-binding cassette, subfamily B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 4; ATP-binding cassette, subfamily B, member 1A; ATP-binding cassette, subfamily B, member 1B; ATP-dependent translocase ABCB1; ATP-dependent translocase ABCB1; multidrug resistance protein 1; bovine p-glycoprotein; CD243; CLCS; colchicin sensitivity; doxorubicin resistance; ecotropic viral integration site 32; Evi32; GP170; mdr; Mdr1; MDR1A; Mdr1b; Mdr3; mdr-3; multi-drug resistance 3; multidrug resistance p-glycoprotein; multidrug resistance protein 1; Multidrug resistance protein 1A; multidrug resistance protein 1B; Multidrug resistance protein 2; multidrug resistance protein 3; multidrug resistance protein MDR1; multiple drug resistant 1a; P glycoprotein 1; P glycoprotein 3; P-glycoprotein; P-glycoprotein 1; P-glycoprotein 2; P-glycoprotein 3; P-glycoprotein/multidrug resistance 1; Pgp; p-gp; PGP1; PGP2; PGY1; Pgy-1; Pgy1-1; PGY2; Pgy3; Pgy-3; Phospholipid transporter ABCB1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170913, 18669, 18671, 24646, 5243 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P06795, P08183, P21447 | |
| ABCB1, Abcb1a, Abcb1b | |
| Synthetic peptide within human P Glycoprotein aa 733-756/1,280. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction