missing translation for 'onlineSavingsMsg'
Learn More

p-Fluorotoluene, 97%, Thermo Scientific Chemicals

Product Code. 10021040
Click to view available options
Quantity:
10 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-32-9
MDL Number MFCD00000358
InChI Key WRWPPGUCZBJXKX-UHFFFAOYSA-N
Synonym 4-fluorotoluene, p-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethylbenzene, toluene, p-fluoro, 1-methyl-4-fluorobenzene, p-fluoromethyl benzene, unii-nud6wz1b8f, p-tolyl fluoride, para-fluorotoluene
PubChem CID 9603
IUPAC Name 1-fluoro-4-methylbenzene
SMILES CC1=CC=C(C=C1)F

Specifications

Color Colorless to Yellow
Density 1.0000g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H7F
Linear Formula CH3C6H4F
Refractive Index 1.4674 to 1.4694 (20°C, 589nm)
Quantity 100 g
Beilstein 05, 290
Merck Index 15, 4212
Molecular Weight (g/mol) 110.131
Formula Weight 110.13
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material p-Fluorotoluene, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Fluorotoluene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.,Highly flammable liquid and vapour.
  • H335-May cause respiratory irritation.,May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/physician/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'