missing translation for 'onlineSavingsMsg'
Learn More

p-Fluorobenzyl Chloride, TRC

Product Code. 30394701
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394701 10 g
30397442 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394701 Supplier Toronto Research Chemicals Supplier No. TRCF58836010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-11-4
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.57
Synonym alpha-Chloro-4-fluorotoluene, alpha-Chloro-p-fluorotoluene, 1-(Chloromethyl)-4-fluorobenzene, 4-Fluorobenzyl Chloride, 4-Fluorophenylmethyl Chloride, NSC 25084, 1-(Chloromethyl)-4-fluorobenzene
IUPAC Name 1-(chloromethyl)-4-fluorobenzene
SMILES Fc1ccc(CCl)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H6ClF/c8-5-6-1-3-7(9)4-2-6/h1-4H,5H2
Formula Weight 144.0142
Chemical Name or Material p-Fluorobenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.