missing translation for 'onlineSavingsMsg'
Learn More

p-Fluorobenzyl Bromide,MP Biomedicals

Product Code. 11412415 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412415 10 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11412415 Supplier MP Biomedicals Supplier No. 0520617205
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 459-46-1
Molecular Formula C7H6BrF
MDL Number MFCD00000359
Quantity 10 g
Synonym 4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide
InChI Key NVNPLEPBDPJYRZ-UHFFFAOYSA-N
SMILES FC1=CC=C(CBr)C=C1
IUPAC Name 1-(bromomethyl)-4-fluorobenzene
Molecular Weight (g/mol) 189.03
PubChem CID 68021
Formula Weight 189.03
Chemical Name or Material p-Fluorobenzyl Bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.