missing translation for 'onlineSavingsMsg'
Learn More

p-Fluoro-Alpha-acetamidocinnamic Acid, TRC

Product Code. 30392851
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392851 1 g
30397222 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392851 Supplier Toronto Research Chemicals Supplier No. TRCF5870001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111649-72-0
Molecular Formula C11 H10 F N O3
Molecular Weight (g/mol) 223.2
Synonym P-Fluoro-A-Acetamidocinnamic Acid
IUPAC Name (E)-2-acetamido-3-(4-fluorophenyl)prop-2-enoic acid
SMILES CC(=O)N\C(=C\c1ccc(F)cc1)\C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H10FNO3/c1-7(14)13-10(11(15)16)6-8-2-4-9(12)5-3-8/h2-6H,1H3,(H,13,14)(H,15,16)/b10-6+
Formula Weight 223.0645
Chemical Name or Material p-Fluoro-a-acetamidocinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.