missing translation for 'onlineSavingsMsg'
Learn More

p-Ethylphenol-d9, TRC

Product Code. 30382692
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382692 100 mg
30371024 50 mg
30371677 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30382692 Supplier Toronto Research Chemicals Supplier No. TRCE925732100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2241872-74-0
Molecular Formula C8HD9O
Molecular Weight (g/mol) 131.22
Synonym Phen-2,3,5,6-d4-ol, 4-(ethyl-1,1,2,2,2-d5)- (ACI), 4-(Ethyl-1,1,2,2,2-d5)phen-2,3,5,6-d4-ol (ACI), 4-Ethylphenol-d9, 4-Ethylphenol-D9, p-Ethylphenol-d9
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1O)C([2H])([2H])C([2H])([2H])[2H]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H10O/c1-2-7-3-5-8(9)6-4-7/h3-6,9H,2H2,1H3/i1D3,2D2,3D,4D,5D,6D
Formula Weight 131.1297
Chemical Name or Material p-Ethylphenol-d9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.