missing translation for 'onlineSavingsMsg'
Learn More

p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals

Catalog Number 10286180
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
40018-26-6
C4H8O2S2
152.23
YUIOPHXTILULQC-UHFFFAOYSA-N
p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme
98330
1,4-dithiane-2,5-diol
C1C(SCC(S1)O)O
This item is not returnable. View return policy
40018-26-6
C4H8O2S2
152.23
YUIOPHXTILULQC-UHFFFAOYSA-N
p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme
98330
1,4-dithiane-2,5-diol
C1C(SCC(S1)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 130.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic bottle
Quantity 250 g
Solubility Information Solubility in water: 3.9g/L (20°C).
Formula Weight 152.23
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material p-Dithiane-2,5-diol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'