missing translation for 'onlineSavingsMsg'
Learn More

p-Dimethylaminobenzaldehyde, For ACS analysis, MP Biomedicals™

Product Code. 11472822 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472822 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11472822 Supplier MP Biomedicals Supplier No. 0215251710
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 100-10-7
Color White to Cream
Molecular Formula C9H11NO
MDL Number MFCD00003381
Quantity 10 g
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
SMILES CN(C)C1=CC=C(C=O)C=C1
IUPAC Name 4-(dimethylamino)benzaldehyde
Molecular Weight (g/mol) 149.19
PubChem CID 7479
Formula Weight 149.2
Physical Form Crystalline Powder
Chemical Name or Material p-Dimethylaminobenzaldehyde, For ACS analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.