missing translation for 'onlineSavingsMsg'
Learn More

p-Cresol, TRC

Product Code. 30134885
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134885 2.5 g 2.5g
18051950 10 g 10g
18070350 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134885 Supplier Toronto Research Chemicals Supplier No. TRCC7819002.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-44-5
Molecular Formula C7 H8 O
Molecular Weight (g/mol) 108.14
Synonym 4-Methylphenol, p-Cresol, Phenol, 4-methyl-, p-Cresol (8CI), 4-Methylphenol, 1-Hydroxy-4-methylbenzene, 1-Methyl-4-hydroxybenzene, 4-Cresol, 4-Hydroxytoluene, NSC 3696, p-Cresylic acid, p-Hydroxytoluene, p-Methylhydroxybenzene, p-Methylphenol, p-Oxytoluene, p-Toluol, p-Tolyl alcohol
IUPAC Name 4-methylphenol
SMILES Cc1ccc(O)cc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H8O/c1-6-2-4-7(8)5-3-6/h2-5,8H,1H3
Formula Weight 108.0575
Percent Purity >95%
Chemical Name or Material 4-Methylphenol (p-Cresol)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.