missing translation for 'onlineSavingsMsg'
Learn More

p-Cresol, Puriss p.a., ≥99.0% (GC),Solstice

Product Code. 15646010
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15646010 500 g Glass bottle 500g
15636010 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15646010 Supplier Honeywell Riedel-de Haën Supplier No. 61030500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 106-44-5
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002376
InChI Key IWDCLRJOBJJRNH-UHFFFAOYSA-N
Synonym p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
PubChem CID 2879
ChEBI CHEBI:17847
IUPAC Name 4-methylphenol
SMILES CC1=CC=C(C=C1)O

Specifications

Melting Point 32°C to 35°C
Density 1.034g/mL at 25°C
Boiling Point 202°C
Flash Point 85°C (185°F)
Ignition Point 1038°F
Packaging Glass bottle
Linear Formula CH3C6H4OH
Quantity 500 g
UN Number UN3455
Beilstein 1305151
Vapor Density 3.72 (vs air)
Formula Weight 108.14g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material p-Cresol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.