missing translation for 'onlineSavingsMsg'
Learn More

p-Cresol, 99%

Product Code. 11458690
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
5000 g
Unit Size:
1000g
100g
250g
5000g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458690 100 g 100g
11435643 250 g 250g
11425643 1000 g 1000g
11445643 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11458690 Supplier Thermo Scientific Alfa Aesar Supplier No. A13531.22
 Restricted Item
In Stock
Add to basket
Ajouter au panier
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 106-44-5 | C7H8O | 108.14 g/mol

p-Cresol is a starting material in the synthesis of bupranolol which is a non-selective beta blocker. It is also used in the production of antioxidants like butylated hydroxytoluene. Furthermore, it is used as an adhesive and a sealant in chemicals, dyes, intermediates, odor agents, plasticizers, plating agents and surface treating agents. It acts as an antioxidant and a disinfectant in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Cresol is a starting material in the synthesis of bupranolol which is a non-selective beta blocker. It is also used in the production of antioxidants like butylated hydroxytoluene. Furthermore, it is used as an adhesive and a sealant in chemicals, dyes, intermediates, odor agents, plasticizers, plating agents and surface treating agents. It acts as an antioxidant and a disinfectant in pharmaceuticals.

Solubility
Soluble in ethanol, diethyl ether and acetone. Slightly soluble in water.

Notes
Air and light sensitive. Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 106-44-5
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002376
InChI Key IWDCLRJOBJJRNH-UHFFFAOYSA-N
Synonym p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
PubChem CID 2879
ChEBI CHEBI:17847
IUPAC Name 4-methylphenol
SMILES CC1=CC=C(C=C1)O

Spécification

Melting Point 32°C to 34°C
Density 1.034
Boiling Point 201°C to 202°C
Flash Point 89°C (192°F)
Odor Phenol-like
Quantity 100 g
UN Number UN3455
Beilstein 1305151
Sensitivity Light sensitive
Merck Index 14,2579
Solubility Information Soluble in ethanol,diethyl ether and acetone. Slightly soluble in water.
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material p-Cresol
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • p-Cresol
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Mention de danger
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contain: 4-methylphenol

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.