missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ P-cadherin Recombinant Rabbit Monoclonal Antibody (K01_1W36)
GREENER_CHOICE

Product Code. 30826953
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826953 100 μL
30827440 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826953 Supplier Invitrogen™ Supplier No. MA563337

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

P-cadherin Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

P-cadherin, also known as Cadherin-3, is a Type 1 cadherin protein that belongs to the cadherin superfamily. Type 1 cadherins are single-pass transmembrane proteins that have 5 extracellular cadherin repeats and an intracellular domain that binds p120-catenin and beta-catenin. Cadherins are calcium-dependent cell-cell adhesion glycoproteins responsible for a range of processes including development, wound healing, cell-cell signaling, cell growth and differentiation. P-cadherin is expressed in human fetal structures, and in adult tissues such as the basal layer of the epidermis, breast, prostate, ovary, cervix, hair follicle, and corneal endothelium. P-cadherin expression has also been reported on embryonic stem cells, and stem cells of the normal mammary gland, and hair follicle. Overexpression of P-cadherin has been associated with poor prognosis in breast, prostate, ovary, colon, and stomach carcinomas. Mutations in CDH3 are associated with congential hypotrichosis with juvenile macular dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P-cadherin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1W36
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene Cdh3
Gene Accession No. P10287, P22223
Gene Alias AI385538; CADH3; cadherin 15, M-cadherin (myotubule); cadherin 3; cadherin 3, type 1, P-cadherin (placental); cadherin-3; Cadp; calcium-dependent adhesion protein, placental; CDH3; CDHP; HJMD; PCAD; P-cadherin; Placental cadherin; Placental-cadherin; RPE-specific cadherin
Gene Symbols Cdh3
Host Species Rabbit
Immunogen A synthetic peptide of human pan Cadherin.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1001, 116777, 12560
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.