missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress (-)-p-Bromotetramisole oxalate

Product Code. p-200104497
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467848 10 mg 10mg
16423676 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16467848 Supplier MedChemExpress Supplier No. HY1969510mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (-)-p-Bromotetramisole Oxalate is a potent and non-specific alkaline phosphatase inhibitor.

Specifications

Product Type Biochemical Assay Reagents
CAS 62284-79-1
Synonym L-p-Bromotetramisole oxalate ∣ 6-Bromolevamisole oxalate
Solubility DMSO : ≥ 29 mg/mL (77.70 mM)
Content And Storage 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Shipping Condition Room temperature
Quantity 10 mg
Molecular Weight (g/mol) 373.22 g/mol
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.