missing translation for 'onlineSavingsMsg'
Learn More

p-Benzoquinone, TRC

Product Code. 30137578
Change view
Click to view available options
Quantity:
1 g
50 g
100 g
Unit Size:
100g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137578 50 g 50g
30137382 1 g 1g
30008365 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137578 Supplier Toronto Research Chemicals Supplier No. TRCB20658050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-51-4
Molecular Formula C6 H4 O2
Molecular Weight (g/mol) 108.09
Synonym p-Benzoquinone (8CI), 1,4-Benzoquinone, 1,4-Cyclohexadienedione, Chinone, Inhibitor PBQ, NSC 36324, PBQ, PBQ 2, Quinone, Stearer PBQ, p-Quinone
IUPAC Name cyclohexa-2,5-diene-1,4-dione
SMILES O=C1C=CC(=O)C=C1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H4O2/c7-5-1-2-6(8)4-3-5/h1-4H
Formula Weight 108.0211
Percent Purity >95%
Chemical Name or Material 1,4-Benzoquinone
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.