missing translation for 'onlineSavingsMsg'
Learn More

p-Arsanilic acid, 98%

Product Code. 11440448
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11440448

Brand: Thermo Scientific Alfa Aesar B24595.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 98-50-0 | C6H8AsNO3 | 217.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-50-0
Molecular Formula C6H8AsNO3
Molecular Weight (g/mol) 217.056
MDL Number MFCD00007819
InChI Key XKNKHVGWJDPIRJ-UHFFFAOYSA-N
Synonym p-arsanilic acid, arsanilic acid, 4-arsanilic acid, 4-aminophenyl arsonic acid, 4-aminobenzenearsonic acid, atoxylic acid, p-aminobenzenearsonic acid, aminophenylarsine acid, p-anilinearsonic acid, pro-gen
PubChem CID 7389
ChEBI CHEBI:49477
IUPAC Name (4-aminophenyl)arsonic acid
SMILES C1=CC(=CC=C1N)[As](=O)(O)O

Specifications

Melting Point >300°C
Quantity 100 g
UN Number UN3465
Beilstein 1102334
Merck Index 14,792
Formula Weight 217.06
Percent Purity 98%
Chemical Name or Material p-Arsanilic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.