missing translation for 'onlineSavingsMsg'
Learn More

p-Anisoyl chloride, 99%

Product Code. 10529440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10529440 500 g 500g
10254270 5 g 5g
10386350 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10529440 Supplier Thermo Scientific Acros Supplier No. 104855000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-07-2 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-07-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000687
InChI Key MXMOTZIXVICDSD-UHFFFAOYSA-N
Synonym p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride
PubChem CID 7477
IUPAC Name 4-methoxybenzoyl chloride
SMILES COC1=CC=C(C=C1)C(Cl)=O

Specifications

Color Colorless to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OC6H4COCl
Refractive Index 1.5800 to 1.5820 (20°C, 589nm)
Quantity 500 g
Beilstein 10, 163
Merck Index 15, 665
Formula Weight 170.6
Percent Purity ≥98.5%
Physical Form Liquid After Melting
Chemical Name or Material p-Anisoyl chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • p-Anisoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.