missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidine, 99%

Product Code. 10030700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10030700 1 kg Glass bottle 1kg
10446900 5 g Glass bottle 5g
10763631 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10030700 Supplier Thermo Scientific Acros Supplier No. 104830010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-94-9 | C7H9NO | 123.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-94-9 , 90-04-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007864
InChI Key BHAAPTBBJKJZER-UHFFFAOYSA-N
Synonym p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine
PubChem CID 7732
ChEBI CHEBI:82388
IUPAC Name 4-methoxyaniline
SMILES COC1=CC=C(C=C1)N

Specifications

Melting Point 56.0°C to 59.0°C
Color Brown to Gray
Density 1.0600g/mL
Boiling Point 240.0°C to 243.0°C
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4NH2
Quantity 1 kg
Beilstein 13, 435
Merck Index 14, 667
Specific Gravity 1.06
Solubility Information Solubility in water: 21g/L (20°C). Other solubilities: freely soluble in methanol, ethanol and ether, soluble in acetone and benzene
Formula Weight 123.15
Percent Purity 99%
Physical Form Crystalline Powder, Granules or Flakes
Chemical Name or Material p-Anisidine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.