Learn More
p-Anisidine, 99%, Thermo Scientific Chemicals
£23.50 - £97.50
Chemical Identifiers
CAS | 104-94-9 |
---|---|
Molecular Formula | C7H9NO |
Molecular Weight (g/mol) | 123.155 |
MDL Number | MFCD00007864 |
InChI Key | BHAAPTBBJKJZER-UHFFFAOYSA-N |
Synonym | p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine |
PubChem CID | 7732 |
ChEBI | CHEBI:82388 |
IUPAC Name | 4-methoxyaniline |
SMILES | COC1=CC=C(C=C1)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488690
|
Thermo Scientific Alfa Aesar
A10946.22 |
100 g |
£23.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498690
|
Thermo Scientific Alfa Aesar
A10946.36 |
500 g |
£58.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494363
|
Thermo Scientific Alfa Aesar
A10946.0B |
1000 g |
£97.50
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.
Solubility
Soluble in acetone, benzene, ether and ethanol. Insoluble in water.
Notes
Air and light sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and chloroformates.
Chemical Identifiers
104-94-9 | |
123.155 | |
BHAAPTBBJKJZER-UHFFFAOYSA-N | |
7732 | |
4-methoxyaniline |
C7H9NO | |
MFCD00007864 | |
p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine | |
CHEBI:82388 | |
COC1=CC=C(C=C1)N |
Specifications
104-94-9 | |
1.089 | |
122°C (251°F) | |
MFCD00007864 | |
471556 | |
p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine | |
BHAAPTBBJKJZER-UHFFFAOYSA-N | |
4-methoxyaniline | |
7732 | |
123.16 | |
p-Anisidine |
56°C to 60°C | |
240°C to 243°C | |
C7H9NO | |
UN2811 | |
14,667 | |
Soluble in acetone,benzene,ether and ethanol. Insoluble in water. | |
COC1=CC=C(C=C1)N | |
123.155 | |
CHEBI:82388 | |
99% |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P262-P264b-P270-P271-P281-P284-P301+P310-P302+P350-P304+P340-P308+P313-P310-P330-P361-P363-P501c
H300+H310+H330-H350-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 203-254-2
missing translation for 'rtecsNumber' : BZ5450000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only